missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HGS Antibody (OTI3H7), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71418H
This item is not returnable.
View return policy
Description
HGS Monoclonal specifically detects HGS in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| HGS | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| HGS | |
| Full length recombinant protein of human HGS (NP_004703) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Western Blot | |
| OTI3H7 | |
| Western Blot | |
| FLJ39878, hepatocyte growth factor-regulated tyrosine kinase substrate, Hrs, HRSFLJ11421, human growth factor-regulated tyrosine kinase substrate, Protein pp110, Vps27, ZFYVE8 | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication, Phospho Specific, Signal Transduction | |
| 9146.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction