missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HGS Monoclonal Antibody (OTI3H7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505894S
This item is not returnable.
View return policy
Description
HGS Monoclonal antibody specifically detects HGS in Canine, Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western BlotSpecifications
| HGS | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q5SW28, Q8WYR1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 23533, 320207, 497931, 612805 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunocytochemistry, Western Blot | |
| OTI3H7 | |
| Unconjugated | |
| PIK3R5 | |
| hepatocyte growth factor-regulated tyrosine kinase substrate; HGF-regulated tyrosine kinase substrate; HGNC:4897; Hgr; Hgs; Hrs; Hrs2; human growth factor-regulated tyrosine kinase substrate; protein pp110; SNAP-25-interacting protein Hrs-2; ZFYVE8 | |
| Full length human recombinant protein of human PIK3R5 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction