missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HHATL Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81952F
This item is not returnable.
View return policy
Description
HHATL Polyclonal antibody specifically detects HHATL in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| HHATL | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| FITC | |
| C3orf3, chromosome 3 open reading frame 3, glycerol uptake/transporter homolog, GUP1, hedgehog acyltransferase-like, Hedgehog acyltransferase-like protein, KIAA1173GUP1, glycerol uptake/transporter homolog (yeast), MBOAT3, membrane bound O-acyltransferase domain containing 3, protein-cysteine N-palmitoyltransferase HHAT-like protein | |
| HHATL antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human HHATL. The immunogen is located within the first 50 amino acids of HHATL. Amino Acid Squence: EASQDGAHRKAFRESVRPG | |
| 0.1 mL | |
| Stem Cells | |
| 57467 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction