missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HHEX Antibody (OTI3E6), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70890AF350
This item is not returnable.
View return policy
Description
HHEX Monoclonal specifically detects HHEX in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| HHEX | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| HHEX | |
| Recombinant fragment expressed in E.coli corresponding to amino acids 100-270 of human HHEX. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2b |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI3E6 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| hematopoietically expressed homeobox, HEXHMPH, Homeobox protein HEX, Homeobox protein PRH, HOX11L-PEN, PRHhematopoietically-expressed homeobox protein HHEX, PRHXhomeobox, hematopoietically expressed, proline-rich homeodomain-containing transcription factor | |
| Mouse | |
| Affinity Purified | |
| Developmental Biology | |
| 3087.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction