missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HHIP Polyclonal Antibody

Description
Recommended positive controls: mouse brain. Predicted reactivity: Mouse (99%), Rat (98%), Xenopus laevis (83%), Rhesus Monkey (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a protein similar to the mouse hedgehog-interacting protein, a regulatory component of the hedgehog signaling pathway. Members of the hedgehog family are evolutionarily conserved proteins which are involved in many fundamental processes in embryonic development, including anteroposterior patterns of limbs and regulation of left-right asymmetry.
Specifications
Specifications
| Antigen | HHIP |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1.33 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 20% glycerol and 0.025% ProClin 300; pH 7 |
| Gene | HHIP |
| Gene Accession No. | Q7TN16, Q96QV1 |
| Gene Alias | hedgehog interacting protein; Hedgehog-interacting protein; Hedgehog-interacting protein-like; Hhip; Hhip1; HIP; RGD1564108; UNQ5825/PRO19644 |
| Gene Symbols | HHIP |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?