missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HHLA3 Monoclonal Antibody (OTI5D12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811165S
This item is not returnable.
View return policy
Description
HHLA3 Monoclonal antibody specifically detects HHLA3 in Human samples. It is validated for Western BlotSpecifications
| HHLA3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O15297 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 8493 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI5D12 | |
| Unconjugated | |
| PPM1D | |
| HERV-H LTR-associating 3; HERV-H LTR-associating protein 3; HHLA3; human endogenous retrovirus-H long terminal repeat-associating 3; human endogenous retrovirus-H long terminal repeat-associating protein 3 | |
| Human recombinant protein fragment corresponding to amino acids 1-375 of human PPM1D produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction