Learn More
Invitrogen™ HHV-6 Early Antigen Monoclonal Antibody (D35B)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA17046
Description
MA1-7046 detects the Human Herpes Virus, type 6. This antibody is specific for a 37 kDa early antigen of HHV6. MA1-7046 has been successfully used in Western blot, immunoprecipitation and immunofluorescence procedures. The MA1-7046 immunogen is Human Herpes Virus type 6 lysate.
Specifications
| HHV-6 Early Antigen | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 |
| Immunoprecipitation, Western Blot, Immunocytochemistry | |
| D35B | |
| Unconjugated | |
| HHV-6; HHV-6 EA | |
| Human Herpes Virus type 6 lysate. | |
| 1 mL | |
| Primary | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.