missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HHV-8 Monoclonal Antibody (LN35)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA126628
This item is not returnable.
View return policy
Description
Store as a concentrated solution. Centrifuge briefly prior to opening vial. Store at 4°C.
HHV8 has been found to be associated with three different diseases observed in AIDS patients; kaposi's sarcoma, primary effusion lymphoma (which is a rare type of non-Hodgkin lymphoma affecting the body cavities) and multicentric Castleman's disease. To date there is much evidence to support a direct role for HHV8 in kaposi's sarcoma.
Specifications
| HHV-8 | |
| Monoclonal | |
| Conc. Not Determined | |
| Tissue culture supernatant with 0.09% sodium azide | |
| Rat | |
| IgG fraction | |
| RUO | |
| Virus | |
| Antibody | |
| IgG2c |
| Immunocytochemistry | |
| LN35 | |
| Unconjugated | |
| HHV8; Human herpesvirus 8; Kaposi's sarcoma-associated herpesvirus; KSHV | |
| Recombinant protein corresponding to the latent nuclear antigen-1 molecule of HHV8. | |
| 500 μL | |
| Primary | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction