missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIC1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560343
This item is not returnable.
View return policy
Description
Immunogen sequence: ELLAQTTHFL HDPKVALESL YPLAKFTAEL GLSPDKAAEV LSQGAHLAAG PDGRTIDRFS PT Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 97%.
Hypermethylated in cancer (HIC-1) was originally identified as a target of p53-induced gene expression. HIC-1 is deleted in the genetic disorder Miller-Dieker syndrome (MDS), and the expression of HIC-1 is also frequently suppressed in leukemia and various cancers due to the hypermethylation of specific DNA regions and the resulting transcriptional silencing. These and other studies indicate that HIC-1 acts as a putative tumor suppressor protein that mediates transcriptional repression. HIC-1 is ubiquitously expressed in adult tissues and its structure is defined by five zinc fingers and an N-terminal broad complex POZ (or BTB) domain. In several BTB/POZ containing proteins, including BCL-6 and the promyelocytic leukemia zinc-finger (PLZF) oncoprotein, this domain interacts with the SMRT/N-CoR-mSin3A HDAC complex and is directly involved in repressing and silencing gene transcription. When this domain is deleted, as with the oncogenic PLZF-RAR chimera of promyelocytic leukemias, this transcriptional repression is attenuated. Conversely, HIC-1 does not interact with components of the HDAC complex, suggesting that HIC-1-induced transcriptional repression is unassociated with the POZ/BTB domain.
Specifications
| HIC1 | |
| Polyclonal | |
| Unconjugated | |
| Hic1 | |
| AA408311; HIC1; hic-1; hypermethylated in cancer 1; hypermethylated in cancer 1 protein; ZBTB29; Zinc finger and BTB domain-containing protein 29; ZNF901 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 3090 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q14526 | |
| Hic1 | |
| Recombinant protein corresponding to Human HIC1. Recombinant protein control fragment (Product #RP-99937). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction