Learn More
HIC2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-81710
 View more versions of this product
Product Code. 18757403
  • € 572.00 / 0.10mL
Description
HIC2 Polyclonal specifically detects HIC2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.