missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HID1 Antibody (OTI2F4), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72481APC
This item is not returnable.
View return policy
Description
HID1 Monoclonal specifically detects HID1 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry.
Specifications
| HID1 | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| HID1 | |
| Full length recombinant protein of human HID1 (NP_085133) produced in HEK293T cells. | |
| 0.1 mL | |
| 283987.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI2F4 | |
| Flow Cytometry | |
| C17orf28, chromosome 17 open reading frame 28, DMC1, DMC1FLJ43526, downregulated in multiple cancer 1, Down-regulated in multiple cancers 1, HID-1, hypothetical protein LOC283987 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction