missing translation for 'onlineSavingsMsg'
Learn More

HID1 Antibody (OTI2F4), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-72481MFV500

Product Code. 18497657

  • € 830.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

HID1 Monoclonal antibody specifically detects HID1 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HID1
Monoclonal
mFluor Violet 500 SE
50mM Sodium Borate
Mouse
Immunogen affinity purified
RUO
283987
Store at 4°C in the dark.
IgG1
Flow Cytometry
OTI2F4
Flow Cytometry
C17orf28, chromosome 17 open reading frame 28, DMC1, DMC1FLJ43526, downregulated in multiple cancer 1, Down-regulated in multiple cancers 1, HID-1, hypothetical protein LOC283987
Full length human recombinant protein of human C17orf28 (NP_085133) produced in HEK293T cell.
0.1 mL
Primary
Human, Mouse, Rat, Canine, Monkey
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.