missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIF-1 alpha Antibody (HA111), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-296PCP
This item is not returnable.
View return policy
Description
HIF-1 alpha Monoclonal antibody specifically detects HIF-1 alpha in Human, Mouse, Rabbit samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| HIF-1 alpha | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rabbit | |
| Purified |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| HA111 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin | |
| AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hypoxia inducible factor 1 alpha subunit, hypoxia inducible factor 1 subunit alpha, hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor), hypoxia-inducible factor 1-alpha, Member of PAS protein 1, member of PAS superfamily 1, MOP1, PAS domain-containing protein 8, PASD8 | |
| This HIF-1 alpha Antibody (HA111) was developed against Human HIF-1 alpha, corresponding to amino acids 329 - 530 [Uniprot# Q16665]. | |
| 0.1 mL | |
| Adaptive Immunity, Angiogenesis, Apoptosis, Autophagy, Cancer, Cardiovascular Biology, Cell Biology, Cell Cycle and Replication, Cellular Markers, Chromatin Research, Cytokine Research, HIF Target Genes, Hypoxia, Hypoxia Inducible Factors (HIF), Hypoxia Signaling, Immunology, Inflammation, Innate Immunity, Lipid and Metabolism, mTOR Pathway, Signal Transduction, Transcription Factors and Regulators | |
| 3091 | |
| Store at 4C in the dark. | |
| IgG2 α |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction