missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ HIF-1 alpha Transcription Factor Assay Kit (Colorimetric)
Assay Kit (Colorimetric)
Brand: Novus Biologicals™ KA1321
797.60 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description

Specifications
Transcription Factor Assay Kit (Colorimetric) | |
Functional, Quantification | |
HIF1A | |
Angiogenesis, Autophagy, Cancer, Cellular Markers, Chromatin Research, HIF Target Genes, Hypoxia, mTOR Pathway, Transcription Factors and Regulators | |
AINT, anti-HIF-1 alpha, anti-HIF1A, ARNT interacting protein, ARNT-interacting protein, Basic-helix-loop-helix-PAS protein MOP1, BHLHE78, Class E basic helix-loop-helix protein 78, HIF1, HIF-1 alpha, HIF1A, HIF-1A, HIF-1alpha, HIF-1-alpha, HIF1-alpha, hypoxia inducible factor 1 alpha subunit, hypoxia inducible factor 1 subunit alpha, hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor), hypoxia-inducible factor 1-alpha, Member of PAS protein 1, member of PAS superfamily 1, MOP1, PAS domain-containing protein 8, PASD8 |
Nuclear Extracts | |
3091 | |
1 Kit | |
Storage of components varies. See protocol for specific instructions. | |
Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction