missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIF-3 alpha Antibody (OTI2D2), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72136IR
This item is not returnable.
View return policy
Description
HIF-3 alpha Monoclonal specifically detects HIF-3 alpha in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| HIF-3 alpha | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HIF3A | |
| Recombinant protein fragment corresponding to amino acids 355-669 of human HIF-3 alpha (NP_690008) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI2D2 | |
| Western Blot, Immunohistochemistry | |
| Basic-helix-loop-helix-PAS protein MOP7, BHLHE17, bHLHe17HIF-3A2, Class E basic helix-loop-helix protein 17, HIF-3A4, HIF-3-alpha, HIF3-alpha, hypoxia inducible factor 3, alpha subunit, hypoxia-inducible factor 3-alpha, hypoxia-inducible factor-3 alpha 4, Inhibitory PAS domain protein, IPASHIF3-alpha-1, Member of PAS protein 7, MOP7PAS domain-containing protein 7, PASD7HIF-3A | |
| Mouse | |
| Affinity Purified | |
| Angiogenesis, Cancer, Chromatin Research, HIF Target Genes, Hypoxia, Transcription Factors and Regulators | |
| 64344.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction