missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HIGD2A Polyclonal Antibody
GREENER_CHOICE

Product Code. 15547182
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15547182 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15547182 Supplier Invitrogen™ Supplier No. PA546477

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: EGLSPTVYRN PESFKEKFVR KTRENPVVPI GCLATAAALT YGLYSFHRGN.

HIGD2A is a proposed subunit of cytochrome c oxidase (COX, complex IV), which is the terminal component of the mitochondrial respiratory chain that catalyzes the reduction of oxygen to water. It may be involved in cytochrome c oxidase activity. It may play a role in the assembly of respiratory supercomplexes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIGD2A
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene HIGD2A
Gene Accession No. Q9BW72
Gene Alias 2010110M21Rik; AA690237; HIG1 domain family member 2A; HIG1 domain family member 2A, mitochondrial; HIG1 domain family, member 2A; HIG1 hypoxia inducible domain family member 2A; HIG1 hypoxia inducible domain family, member 2A; Higd2a; RCF1 homolog B; RCF1b
Gene Symbols HIGD2A
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human HIGD2A (aa 22-71).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 192286
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.