missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIGD2B Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA582657
This item is not returnable.
View return policy
Description
Immunogen sequence: MATLGFVTPE APFESSKPPI FEGLSPTVYS NPEGFKEKFL RKTRENP Highest antigen sequence indentity to the following orthologs: Rat - 74%, Mouse - 79%.
HIGD2B is a protein coding gene. Gene ontology (GO) annotation include integral component of membrane.
Specifications
| HIGD2B | |
| Polyclonal | |
| Unconjugated | |
| HIGD2B | |
| HIG1 hypoxia inducible domain family member 2B; HIGD2B; HIGD2BP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 123346 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q4VC39 | |
| HIGD2B | |
| Recombinant protein corresponding to Human HIGD2B. Recombinant protein control fragment (Product #RP-91011). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction