Learn More
Agilent BioTek High Flow Complete Dispense/Waste System
GREENER_CHOICE
Product Code. p-200007356 Shop All Agilent BioTek Products
Click to view available options
Capacity (Metric):
4 L
10 L
20 L
Unit Size:
Each
Product Code. 17854657
Brand: Agilent BioTek 2120532
High Flow Complete Dispense/Waste System for use with the 406 FX Washer Dispenser.
  • Includes four dispense bottles, two waste bottles and high flow vacuum pump
TRUSTED_SUSTAINABILITY
Specifications
Includes 4 x 4 L Dispense Bottles, 2 x 4 L Waste Bottles, High Flow Vacuum Pump
Capacity (Metric) 4 L
Type Waste System