Learn More
Kartell™ High Impact Polystyrene Balance Boats
Product Code. p-8005827
Click to view available options
Capacity (Metric):
5 mL
7 mL
30 mL
100 mL
250 mL
Color:
Black
White
Shape:
Diamond
Square
Unit Size:
Pack of 1000
Pack of 250
Pack of 500
Product Code. 10447512
Brand: Kartell™ 1110003
Only null left
Add to basket
Add to basket
Provide an easy and inexpensive system for weighing any medium.
  • Lightweight weighing boats
  • Non-contaminating (made of HIPS with hydrophobic surface)
  • Easily fashioned to make funnel shape
  • Available in two models: diamond (DS) and square (SQ) 
  • DS model available in white or black for visibility of trace sample remains adhering to boat
  • Reproducible tare weight: batch to batch variation less than 5%
  • Antistatic
Specifications
Bottom Shape Flat
Capacity (Metric) 5 mL
Color White
Material Polystyrene
Shape Diamond
Type Weighing Boat
Dimensions (L x W x H) 31 x 53 x 5.3 mm
Length (Metric) 31 mm
Width (Metric) 53 mm
Height (Metric) 5.3 mm
Weight (Metric) 0.3 g