Learn More
Masterflex™ High Pressure Tubing with Luer Ends
Description
- Plastic Luer Assemblies
- Get a seamless transition from source to system
- Each assembly comes packaged individually; can be sterilized
- High pressure tubing has luer fittings at each end
- Male luer lock x female luer in varying lengths from 10 to 48 in.
- Sterilize by ETO
Specifications
Specifications
| Material | PVC |
| Includes | 2 ports: 1 Male, 1 female luer lock end |
| Maximum Pressure | 900 psi |
| Product Type | High Pressure tubing with Luer Ends |