Learn More
Aalborg™ High Resolution Valve Brass
Click to view available options
Airflow Rate:
2500 mL/min.
400 mL/min.
Waterflow Rate:
12mL/min.
70mL/min.
Unit Size:
Each
Description
- For high resolution
- Designed for laboratory and industrial applications for regulating corrosive gases and liquids or for high purity service.
- They may also be used as shut off valves.
- Bodies of the valves are reinforced by structurally rigid metallic shells.
- Shells are made of brass, bushings are made of plated brass or 316 stainless steel.
Especificaciones
Especificaciones
| Type | High Resolution Valve |
| Airflow Rate | 400 mL/min. |
| Waterflow Rate | 12mL/min. |
| Max. Operating Temperature (Metric) | 82°C |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido