Learn More
High Temperature Viscosity Standard; UKAS ISO/IEC17025 and ISO 17034 certified, viscosity 149.2 mPa.s (60 °C), Paragon Scientific™
Code produit. 16030280
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
500 mL
missing translation for 'unitSize':
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit. Quantity missing translation for 'unitSize'
16030280 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit. 16030280 missing translation for 'mfr' Paragon Scientific missing translation for 'supplierNo' N600
 Restricted Item
Estimated Shipment: 13-10-2026
Ajouter au panier
Ajouter au panier
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Paragon ISO 17025 / ISO 17034 certified high temperature viscosity standards are for the calibration and verification of glass capillary viscometers and other viscosity measuring equipment where operating temperature is controlled precisely.
  • Certified according to ISO 17025 / ISO 17034 under our UKAS accreditation
  • Kinematic (cSt), dynamic viscosity (mPa·s) and density (g/mL) given at all temperatures
  • Standard Pack Size 500mL
  • Fully traceable to National Standards
  • Ensures full compliance to ASTM & IP test method protocol
  • Tamper evident security packaging
  • 2 Year shelf life
  • Manufactured in the United Kingdom
Spécification
Quantity 500 mL
Nominal values quoted. Certified Values will vary from batch to batch.