missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syngene™ HILED Upgrade Lighting from RGB to RGBIR
Shop All Syngene Products

Click to view available options
Color:
Far Red
Infra Red
Red, Green, Blue, UV
Unit Size:
Each
Description
For use with GBOX
Specifications
Specifications
| For Use With (Application) | Multiplex Western Fluorescence |
| Color | Infra Red |
| For Use With (Equipment) | Gbox Chemi Systems |
| Quantity | 1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction