Learn More
Kinematica™ Hinge Stand ST-P01-200 for Polytron™ PT 1300 D Homogenizer
Product Code. 13457639 Shop All Kinematica Products
Click to view available options
For Use With (Equipment):
PT 1300 D Disperser
Unit Size:
Each
Product Code. 13457639
Brand: Kinematica™ 11040012
Only null left
Add to basket
Add to basket
Stainless steel swivel stand (gas spring) and base mounting. The drive support can be rotated in any direction. It can be raised or lowered.
Joint stand (gas spring) with two rivet plugs stainless steel for use with Polytron™ PT 1300 D drive
Specifications
Product Type Hinge Stand
Material Stainless Steel
For Use With (Equipment) PT 1300 D Disperser
Model ST-P 01/200