Learn More
HINT1 Mouse, anti-Human, CoraLite Plus 488, Clone: 1G3E4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-67583-100UL
 View more versions of this product
Product Code. 18025285
  • € 450.00 / 100µL
Description
The HINT1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HINT1. This antibody recognizes human antigen. The HINT1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
HINT1
1G3E4
CoraLite Plus 488
Mouse
Primary
Human
155 kDa
IgG2a
Immunofluorescence
1000 μg/mL
FLJ30414, FLJ32340, HINT, HINT1, PKCI 1, PKCI1, PRKCNH1, Protein kinase C inhibitor 1
100 μL
3094
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.