missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HINT2 Antibody (AT5B10), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59387IR
This item is not returnable.
View return policy
Description
HINT2 Monoclonal antibody specifically detects HINT2 in Human samples. It is validated for Western Blot, ELISA
Specifications
| HINT2 | |
| Monoclonal | |
| DyLight 755 | |
| EC 3.-, HINT-2, HINT-3, histidine triad nucleotide binding protein 2, histidine triad nucleotide-binding protein 2, mitochondrial, HIT-17, HIT-17kDa, PKCI-1-related HIT protein, protein kinase C inhibitor-2 | |
| Recombinant human HINT2 (18-163aa) purified from E. coli | |
| 0.1 mL | |
| Signal Transduction | |
| 84681 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA | |
| AT5B10 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction