missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIP-55 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-1036
This item is not returnable.
View return policy
Description
HIP-55 Polyclonal specifically detects HIP-55 in Human, Mouse samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| HIP-55 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| ABP1, Cervical SH3P7, CMAP, Drebrin-F, drebrin-like, drebrin-like protein, HIP-55SH3P7Cervical mucin-associated protein, HPK1-interacting protein of 55 kDa, SH3 domain-containing protein 7, src homology 3 domain-containing protein HIP-55 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| This antibody is expected to recognize all reported isoforms (NP_054782.2; NP_001014436.1; NP_001116428.1) | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.03 - 1 ug/ml, Peptide ELISA Detection limit 1:16000 | |
| NP_054782. | |
| DBNL | |
| Peptide with sequence AANLSRNGPALQE-C corresponding to N-Terminus according to NP_001014436.1, NP_001116428.1. | |
| 0.1 mg | |
| Signal Transduction | |
| 28988 | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction