Learn More
HIP-55 Antibody (OTI6C9), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-71841AF647
Product Code. 18168804
  • € 751.00 / 0.10mL
Description
HIP-55 Monoclonal specifically detects HIP-55 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HIP-55
Monoclonal
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
DBNL
Full length recombinant protein of human HIP-55 (NP_054782) produced in HEK293T cells.
0.1 mL
Primary
Human, Mouse, Rat
IgG1
Immunohistochemistry
OTI6C9
Immunohistochemistry
ABP1, Cervical SH3P7, CMAP, Drebrin-F, drebrin-like, drebrin-like protein, HIP-55SH3P7Cervical mucin-associated protein, HPK1-interacting protein of 55 kDa, SH3 domain-containing protein 7, src homology 3 domain-containing protein HIP-55
Mouse
Affinity Purified
Signal Transduction
28988.0
Store at 4°C in the dark.