missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HIP Monoclonal Antibody (K24012_7H12), Invitrogen™

Product Code. p-200088671
Change view
Click to view available options
Quantity:
50 μL
200 μL
Unit Size:
200µg
50µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30199708 50 μL 50µg
30208076 200 μL 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30199708 Supplier Invitrogen™ Supplier No. MA555820

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Sequence of this protein is as follows: MDPRKVNELR AFVKMCKQDP SVLHTEEMRF LREWVESMGG KVPPATQKAK SEENTKEEKP DSKKVEEDLK ADEPSSEESD LEIDKEGVIE PDTDAPQEMG DENAEITEEM MDQANDKKVA AIEALNDGEL QKAIDLFTDA IKLNPRLAIL YAKRASVFVK LQKPNAAIRD CDRAIEINPD SAQPYKWRGK AHRLLGHWEE AAHDLALACK LDYDEDASAM LKEVQPRAQK IAEHRRKYER KREEREIKER IERVKKAREE HERAQREEEA RRQSGAQYGS FPGGFPGGMP GNFPGGMPGM GGGMPGMAGM PGLNEILSDP EVLAAMQDPE VMVAFQDVAQ NPANMSKYQS NPKVMNLISK LSAKFGGQA

The protein encoded by this gene is an adaptor protein that mediates the association of the heat shock proteins HSP70 and HSP90. This protein has been shown to be involved in the assembly process of glucocorticoid receptor, which requires the assistance of multiple molecular chaperones. The expression of this gene is reported to be downregulated in colorectal carcinoma tissue suggesting that is a candidate tumor suppressor gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen HIP
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K24012_7H12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene St13
Gene Accession No. P50502
Gene Alias 1110007I03Rik; 3110002K08Rik; AAG2; aging-associated protein 2; AW555194; FAM10A1; FAM10A4; heat shock 70kD protein binding protein; hedgehog interacting protein; hedgehog-interacting protein; HHIP; HIP; HOP; Hsc70-interacting protein; Hsp70 interacting protein; Hsp70-interacting protein; HSPABP; HSPABP1; hypothetical protein; p48; PRO0786; Progesterone receptor-associated p48 protein; Protein FAM10A1; Protein ST13 homolog; putative tumor suppressor ST13; RCJMB04_6h13; Renal carcinoma antigen NY-REN-33; SNC6; ST13; ST13 Hsp70 interacting protein; ST13, Hsp70 interacting protein; ST13P5; suppression of tumorigenicity 13; suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein); suppression of tumorigenicity 13 (colon carcinoma) (Hsp70 interacting protein) pseudogene 5; suppression of tumorigenicity 13 (colon carcinoma) Hsp70-interacting protein; suppression of tumorigenicity 13 protein; testis secretory sperm-binding protein Li 233m; UNQ5825/PRO19644
Gene Symbols St13
Host Species Mouse
Immunogen Recombinant human ST13 (immunogen range is 1-369).
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6767
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.