Learn More
Aromatase Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-99064APC
Product Code. 18622086
  • € 407.00 / 0.10mL
Description
Aromatase Polyclonal antibody specifically detects Aromatase in Human samples. It is validated for Immunohistochemistry (Paraffin)