missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIPK1 Antibody (OTI5F1), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72460IR
This item is not returnable.
View return policy
Description
HIPK1 Monoclonal specifically detects HIPK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| HIPK1 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| HIPK1 | |
| Recombinant protein fragment corresponding to amino acids 439-697 of human HIPK1 (NP_852003) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| OTI5F1 | |
| Western Blot, Immunohistochemistry | |
| EC 2.7.11.1, homeodomain interacting protein kinase 1, homeodomain interacting protein kinase 1-like protein, homeodomain-interacting protein kinase 1, KIAA0630MGC26642, MGC33446, MGC33548, Myak, Nbak2, nuclear body associated kinase 2b | |
| Mouse | |
| Affinity Purified | |
| Protein Kinase | |
| 204851.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction