missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIPK1 Monoclonal Antibody (OTI3C4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807264S
This item is not returnable.
View return policy
Description
HIPK1 Monoclonal antibody specifically detects HIPK1 in Human samples. It is validated for Western BlotSpecifications
| HIPK1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q86Z02 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 204851 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI3C4 | |
| Unconjugated | |
| HIPK1 | |
| 1110062K04Rik; EMBL:ABO47653.1}; Hipk1; hipk1 {ECO:0000312; homeodomain interacting protein kinase 1; homeodomain interacting protein kinase 1-like protein; homeodomain-interacting protein kinase 1; KIAA0630; MYAK; NBAK2; nuclear body associated kinase 2b; nuclear body-associated kinase 2; Protein kinase Myak | |
| Human recombinant protein fragment corresponding to amino acids 439-697 of human HIPK1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction