missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIPK2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00 - € 326.00
Specifications
| Antigen | HIPK2 |
|---|---|
| Dilution | Immunohistochemistry 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18636865
|
Novus Biologicals
NBP2-99584-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18695445
|
Novus Biologicals
NBP2-99584-50ul |
50 μg |
€ 216.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HIPK2 Polyclonal antibody specifically detects HIPK2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifikationer
| HIPK2 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis | |
| PBS (pH 7.0) | |
| 28996 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunohistochemistry 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| DKFZp686K02111, EC 2.7.11.1, FLJ23711, hHIPk2, homeodomain interacting protein kinase 2, homeodomain-interacting protein kinase 2, PRO0593 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human HIPK2. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel