missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hippocalcin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06199-50ul
This item is not returnable.
View return policy
Description
Hippocalcin Polyclonal antibody specifically detects Hippocalcin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Hippocalcin | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| BDR2, Calcium-binding protein BDR-2, hippocalcin, neuron specific calcium-binding protein hippocalcin, neuron-specific calcium-binding protein hippocalcin | |
| Produced in rabbits immunized with E. coli-derived Human Hippocalcin fragment. | |
| 50 μg | |
| Neuroscience | |
| 3208 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction