missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hippocalcin like 4 Antibody (OTI3C3), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-46002
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Western Blot
Hippocalcin like 4 Monoclonal specifically detects Hippocalcin like 4 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Hippocalcin like 4 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:2000 | |
| hippocalcin like 4, hippocalcin-like protein 4, HLP4DKFZp761G122 | |
| Mouse | |
| 22 kDa | |
| 0.1 mL | |
| Vision | |
| 51440 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot | |
| OTI3C3 | |
| Unconjugated | |
| PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
| HPCAL4 | |
| Human recombinant protein fragment corresponding to amino acids 2-191 of human HPCAL4(NP_057341) produced in E.coli. | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction