missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIRA, Mouse anti-Human, Clone: WC119.2H11, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB8967.100ug
This item is not returnable.
View return policy
Specifications
| HIRA | |
| Monoclonal | |
| 1 μg/μL | |
| The optimal working dilution should be determined by the end user. | |
| HIR histone cell cycle regulation defective homolog A (S. cerevisiae) | |
| HIRA | |
| Recombinant protein corresponding to amino acids 421-729 of human HIRA. | |
| RUO | |
| 7290 | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| WC119.2H11 | |
| Unconjugated | |
| In PBS, pH 7.4 (0.02% sodium azide) | |
| DGCR1/TUP1/TUPLE1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| This antibody reacts with human HIRA. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction