missing translation for 'onlineSavingsMsg'
Learn More

His Tag Alexa Fluor 647-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18737459
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18737459 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18737459 Supplier R&D Systems Supplier No. IC050R100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

His Tag Monoclonal specifically detects His Tag in Multi-Species samples. It is validated for Flow Cytometry, Intracellular Staining by Flow Cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen His Tag
Applications Flow Cytometry
Classification Monoclonal
Clone AD1.1.10
Conjugate Alexa Fluor 647
Dilution Flow Cytometry 0.25 -1.0 ug/10^6 cells, Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Alias 6 His epitope tag, 6X His, 6X-His, H, HHHHHH epitope tag, HHHHHH tag, HIS, His tag, polyHistidine, Poly-histidine
Host Species Mouse
Immunogen His-tagged peptide
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects proteins containing accessible consecutive histidine regions. The antibody detects His tags localized at the amino- or carboxyl-terminus.
Target Species Multi-species
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.