Learn More
Cytiva HiScreen Capto Q ImpRes Prepacked Column
Product Code. 11653074 Shop All Cytiva Products
Change view
Click to view available options
Diameter (Metric) Inner:
7.7 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner unitSize
11653074 7.7 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11653074 Supplier Cytiva Supplier No. 17547015
Powering Proteomics
Use promo code 28003
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
HiScreen Capto Q ImpRes columns are prepacked with Capto Q ImpRes ion exchange chromatography medium, and are part of the process development platform available from Cytiva. The columns are an excellent choice for method optimization and parameter screening.
  • Capto Q ImpRes is a strong anion exchange BioProcess medium, designed to meet the demands of modern large-scale manufacturers for fast, efficient, and cost-effective intermediate and polishing protein purification.
  • 10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening.
  • Easily connected in series to achieve 20 cm bed height.
  • Small bed volume gives fast results and minimal sample/buffer consumption.
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity.
TRUSTED_SUSTAINABILITY
Specifications
Bed Volume 4.7
Diameter (Metric) Inner 7.7 mm
Flow Rate 100 to 300 cm/h
Max. Pressure 4 bar [0.4 MPa] (59 psi)
Content And Storage 4° to 30°C, in 20% Ethanol
Column Type Ion Exchange Column
For Use With (Application) Ion Exchange Chromatography
compliance-icons
Product Identifier
  • HiScreen Capto Q ImpRes, 4.7 ml
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.