Learn More
Cytiva HiScreen Phenyl FF (High Sub)
Product Code. 10353310 Shop All Cytiva Products
Change view
Click to view available options
Diameter (Metric) Inner:
7.7 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner unitSize
10353310 7.7 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10353310 Supplier Cytiva Supplier No. 28926988
HiScreen Phenyl FF (high sub) columns are prepacked with Phenyl Sepharose 6 Fast Flow hydrophobic interaction chromatography (HIC) media. The columns are an excellent choice for method optimization and parameter screening.
  • Phenyl Sepharose 6 Fast Flow is a well established, standard aromatic HIC medium for capture and intermediate purification requiring low to medium hydrophobicity.Two levels of substitution help find optimal selectivity and binding capacity for a given application.
  • Prepacked, ready-to-use columns for convenient process development optimization.
  • 10 cm bed height of HiScreen columns is designed to allow method optimization and parameter screening.
  • Easily connected in series to achieve 20 cm bed height.
  • Small bed volume gives fast results and minimal sample/buffer consumption.
  • Reproducible results, scalable to BioProcess columns packed with the same media using the same linear fluid velocity.
TRUSTED_SUSTAINABILITY
Specifications
Description HiScreen Phenyl FF (High Sub)
Bed Volume 4.7
Diameter (Metric) Inner 7.7 mm
Flow Rate Less than 450 cm/h
Max. Pressure 1.5 bar [0.15 MPa] (22 psi)
Content And Storage 4° to 30°C, 20% Ethanol or 0.01M NaOH
Column Type HIC Column
For Use With (Application) Hydrophobic Interaction Chromatography
compliance-icons
Product Identifier
  • HiScreen Phenyl FF (high sub)
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: ethanol
  • UNKNOWN ACUTE-16.5 per cent of the mixture consists of component(s) of unknown acute toxicity.