Learn More
Cytiva HiScreen™ Q Sepharose™ and SP Sepharose™ Fast Flow IEX Columns
Product Code. p-4595752 Shop All Cytiva Products
Click to view available options
Product Type:
HiScreen Q Sepharose FF Column
HiScreen SP Sepharose FF clumn
Ligand Type:
Quaternary Amine
Sulphopropyl
pH Range:
2 to 12
4 to 13
Unit Size:
Each
Product Code. 10201230
Brand: Cytiva 28950510
Designed for method optimization and parameter screening
  • Ready-to-use column for convenient process development optimization and parameter screening
  • Highly cross-linked agarose matrix with average bead size of 90μm
  • Bed height of 10cm designed to allow method optimization and parameter screening
  • 20cm bed height is acheived when it is connected in series
  • Small bed volume (4.7mL) gives fast result and minimal sample/buffer consumption
  • Reproducible results, scalable to BioProcess columns packed with the same chromatography media using the same linear fluid velocity
  • Bed dimension: 7.7 × 100mm
  • Matrix: 6% cross-linked agarose beads
  • Average particle size: 90μm
  • Max. flow velocity: 450cm/hr.
  • Material (column hardware and column tube): polypropylene
  • Column I.D.: 7.7mm
HiScreen Q FF
  • Media:
  • pH stability working range: 2 to 12
  • pH stability cleaning: 2 to 14
  • Ion exchanger type: strong anion exchanger
HiScreen SP FF
  • Media:
  • pH stability working range: 3 to 14
  • pH stability cleaning: 4 to 13
  • Ion exchanger type: strong cation exchanger
TRUSTED_SUSTAINABILITY
Specifications
Buffer 1M NaOH, 8M Urea, 8M Guanidine Hydrochloride, 70% Ethanol
Product Type HiScreen Q Sepharose FF Column
Bed Volume 4.7mL
Diameter (English) Inner 0.3 in.
Diameter (Metric) Inner 7.7mm
Quantity 1 x 4.7mL
Height (English) 3.93 in.
Height (Metric) 100mm
Flow Rate <450 cm/hr.
Material Polypropylene
Ligand Type Quaternary Amine
Max. Pressure 1.5 bar
pH Range 2 to 12
Particle Size 45 to 165μm
Support Type 6% Cross-linked Agarose
Content And Storage 4° to 30°C
Column Type Ion Exchange Column
For Use With (Application) Optimization and parameter screening, ion exchange chromato graphy
Show More Show Less
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.