missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIST1H2AG Antibody (CL5041), Novus Biologicals™
Mouse Monoclonal Antibody
€ 415.00 - € 572.00
Specifications
| Antigen | HIST1H2AG |
|---|---|
| Clone | CL5041 |
| Dilution | Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18259313
|
Novus Biologicals
NBP2-61152 |
100 μL |
€ 572.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18659536
|
Novus Biologicals
NBP2-61152-25ul |
25 μL |
€ 415.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HIST1H2AG Monoclonal specifically detects HIST1H2AG in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| HIST1H2AG | |
| Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Unconjugated | |
| Mouse | |
| 8969 | |
| This antibody was developed against a synthetic peptide corresponding to the following amino acid sequence:RSSRAGLQFPVGRVHR | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| CL5041 | |
| Monoclonal | |
| Purified | |
| H2A histone family, member P, H2A.1, H2A.1b, H2A/p, H2AFC, H2AFD, H2AFI, H2AFN, H2AFP, HIST1H2AI, HIST1H2AK, HIST1H2AL, HIST1H2AM, histone 1, H2ag, histone cluster 1, H2ag, histone H2A type 1, Histone H2A/p, pH2A/f | |
| HIST1H2AG | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title