Learn More
HIST1H2BC Antibody (CL5101) [Alexa Fluor™ 532], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61154AF532
Product Code. 30304245
  • € 737.00 / 0.10mL
Description
HIST1H2BC Monoclonal antibody specifically detects HIST1H2BC in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HIST1H2BC
Monoclonal
Alexa Fluor 532
H2B histone family, member K, H2B/a, H2B/g, H2B/kH2B/h, H2BFA, H2BFG, H2BFH, H2BFKH2B/l, H2BFL, HIST1H2BC, HIST1H2BE, HIST1H2BF, HIST1H2BG, histone 1, H2bi, histone cluster 1, H2bi, histone H2B type 1-C/E/F/G/I, Histone H2B.1 A, Histone H2B.a, Histone H2B.g, Histone H2B.h, Histone H2B.k, Histone H2B.l
This antibody was developed using a synthetic peptide derived from P62807, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Human, Mouse, Rat
Purified
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL5101
50mM Sodium Borate
Mouse
Protein A purified
RUO
8347
Store at 4⪚C in the dark.