missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIST1H3A, Formyl Lys122 Antibody (32C1), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 343.00 - € 533.00
Specifications
| Antigen | HIST1H3A |
|---|---|
| Clone | 32C1 |
| Dilution | ELISA, Immunohistochemistry 1:50-1:200 |
| Applications | ELISA, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043455
|
Novus Biologicals
NBP3-26574-50ul |
50 μL |
€ 343.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043570
|
Novus Biologicals
NBP3-26574-100ul |
100 μL |
€ 533.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
HIST1H3A Monoclonal antibody specifically detects HIST1H3A in Human samples. It is validated for ELISA, ImmunohistochemistrySpecifications
| HIST1H3A | |
| ELISA, Immunohistochemistry 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| H3 histone family, member A, H3/A, H3FAHIST1H3E, H3FB, H3FC, H3FD, H3FF, H3FH, H3FI, H3FJ, H3FK, H3FL, HIST1H3B, HIST1H3C, HIST1H3D, HIST1H3F, HIST1H3G, HIST1H3H, HIST1H3I, HIST1H3J, histone 1, H3a, histone cluster 1, H3a, histone H3.1, Histone H3/a, Histone H3/b, Histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h, Histone H3/i, Histone H3/j, Histone H3/k, Histone H3/l | |
| A synthesized peptide derived from Human HIST1H3A. The HIST1H3A [Formyl Lys122] Recombinant Monoclonal Antibody was raised against the peptide IMP-(Fo)K-DI formylated at K122. [UniProt P68431] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
| 32C1 | |
| ELISA, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Breast Cancer, Cancer, Cellular Markers, Core ESC Like Genes, Oncogenes, Protein Kinase, Stem Cell Markers, Tumor Suppressors | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 8350 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title