missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIST1H3D Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20532-1-AP-20UL
This item is not returnable.
View return policy
Specifications
| HIST1H3D | |
| Polyclonal | |
| Unconjugated | |
| HIST1H3D | |
| H3/b, H3FA, H3FB, H3FC, H3FD, H3FF, H3FH, H3FI, H3FJ, H3FK, H3FL, HIST1H3A, HIST1H3B, HIST1H3C, HIST1H3D, HIST1H3E, HIST1H3F, HIST1H3G, HIST1H3H, HIST1H3I, HIST1H3J, histone cluster 1, H3d, Histone H3.1, Histone H3/a, Histone H3/b, Histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h, Histone H3/i, Histone H3/j, Histone H3/k, Histone H3/l | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 319149, 8351 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P68431 | |
| Hist1h3d | |
| HIST1H3D Fusion Protein Ag14394 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction