missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIST1H4H, Mouse anti-Human, Clone: 6D4, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00008365-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: TKPAIRRLARRGGVKRISGLIYEETRGVLKVFLENVIRDAVTYTEHAKRKTVTAMDVVYALKRQGRTLYGFGGSpecifications
| HIST1H4H | |
| Monoclonal | |
| Unconjugated | |
| histone cluster 1, H4h | |
| H4/h/H4FH | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence | |
| 6D4 | |
| In 1x PBS, pH 7.4 | |
| NM_003543 | |
| HIST1H4H | |
| HIST1H4H (NP_003534, 31 a.a. ∼ 103 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 8365 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction