missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histamine H1R Antibody, Novus Biologicals™
Goat Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-06039
This item is not returnable.
View return policy
Description
Histamine H1R Polyclonal specifically detects Histamine H1R in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
Specifications
| Histamine H1R | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| H1R, H1-R, hisH1, histamine receptor H1, subclass H1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.5 to 1.5 μg/mL, Simple Western, Immunohistochemistry 2-4μg/mL, Immunohistochemistry-Paraffin 2-4μg/mL, Peptide ELISA Detection limit 1:16000 | |
| NP_000852.1 | |
| HRH1 | |
| Peptide with sequence QQTSVRREDKCETD corresponding to internal region according to NP_000852.1. | |
| 0.1 mg | |
| GPCR, Immune System Diseases, Immunology, Innate Immunity | |
| 3269 | |
| Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction