missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histamine H3R Antibody (2420B) [Janelia Fluor™ 646], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB102001J
This item is not returnable.
View return policy
Description
Histamine H3R Monoclonal antibody specifically detects Histamine H3R in Human, Mouse, Rat samples. It is validated for Flow Cytometry, CyTOF
Specifications
| Histamine H3R | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry, CyTOF | |
| 2420B | |
| Flow Cytometry, CyTOF-ready | |
| GPCR97G protein-coupled receptor 97, G-protein coupled receptor 97, H3R, HH3R, histamine H3 receptor, histamine receptor H3 | |
| Synthetic peptide containing Human/Mouse/Rat Histamine H3 R, Accession # Q9Y5N1 | |
| 0.1 mL | |
| GPCR, Immunology, Innate Immunity | |
| 11255 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction