missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Histamine Monoclonal Antibody (1F3)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA549687
This item is not returnable.
View return policy
Description
Antibody tested in indirect ELISA.
Histamine is located in mast cells, endocrine cells of the gut, blood cells and in some cells of the peripheral and central nervous system. Histamine is a potent vasodilator when secreted by mast cell found in various tissues as a result of allergic hytersensitivity or inflammation. In the central nervous system, Histamine is putative neurotransmitter. In the brain, its highest content has been found in the hypothalamus and in certain areas of the mesencephalon. The Histamine antiserum has a sensitivity level capable of detecting the low level Histamine contents of the brain.
Specifications
| Histamine | |
| Monoclonal | |
| 6.7 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Conjugate of HIS hapten and KLH. | |
| 200 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Lateral Flow, Lateral Flow, SDS-Page | |
| 1F3 | |
| Unconjugated | |
| Mouse | |
| Protein G | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction