Learn More
Histamine N-Methyltransferase/HNMT Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP3-00070JF646
Product Code. 18501340
  • € 407.00 / 0.10mL
Description
Histamine N-Methyltransferase/HNMT Polyclonal antibody specifically detects Histamine N-Methyltransferase/HNMT in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Western Blot, ELISA, Immunoprecipitation
Store at 4C in the dark.