missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Histone (Calf)

Product Code. 16161770
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16161770 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16161770 Supplier Abnova™ Supplier No. P5231.250mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for SDS-PAGE

#REF!

Specifications

For Use With (Application) SDS-PAGE
Name Histone (Calf)
Preparation Method Native protein purified from Calf Thymus
Quantity 250 mg
Storage Requirements Store at 4°C.
Regulatory Status RUO
Common Name HISTONE
Species Bovine
Protein Tag None
Expression System Native protein purified from Calf Thymus
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.